Learn More
Brady™ Marking Tape T-Shaped Divider Marks: Adhesive Vinyl, Solid Color, Black
Permanent acrylic adhesive sticks to a wide variety of surfaces
Manufacturer:  Brady™ 121422
Catalog No. 19-103-859
Add to cart
Description
Description
  • Indoor/outdoor vinyl film withstands exposure to moisture, chemicals and handling.
  • Up to 3 year average outdoor durability
Specifications
Specifications
Roll of 545 Each
No Text
Vinyl
1.1 in.
1.5 in.
Safety
Black
Marking Tape
0.004 in.
Permanent Acrylic