Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ MART-1/Melan-A (Melanoma Marker) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MART-1/Melan-A (Melanoma Marker) Ab-1, Mouse Monoclonal Antibody.

$173.40 - $741.50

Specifications

Antigen MART-1/Melan-A (Melanoma Marker) Ab-1
Clone M2-7C10
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 8
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS612P
View Documents
Thermo Scientific™
MS612P
Purified Ab with BSA and Azide 1mL
Each for $703.50
Add to cart
 
MS612P0
View Documents
Thermo Scientific™
MS612P0
Purified Ab with BSA and Azide 100μL
Each for $173.40
Add to cart
 
MS612P1
View Documents
Thermo Scientific™
MS612P1
Purified Ab with BSA and Azide 500μL
Each for $464.00
Add to cart
 
MS612P1ABX
View Documents
Thermo Scientific™
MS612P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $518.50
Add to cart
 
MS612PABX
View Documents
Thermo Scientific™
MS612PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $741.50
Add to cart
 
MS612R7
View Documents
Thermo Scientific™
MS612R7
Ready-to-use for IHC (UVLP) 7mL
Each for $418.00
Add to cart
 
MS612RQ
View Documents
Thermo Scientific™
MS612RQ
Ready-to-use for IHC (Quanto) 12mL
Each for $455.00
Add to cart
 
MS612R1
View Documents
Thermo Scientific™
MS612R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MART-1/Melan-A (Melanoma Marker) Ab-1, Mouse Monoclonal Antibody.

MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-Ais a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum.

Host Species: Mouse

Clone: M2-7C10

Isotype: IgG2b/κ

Species Reactivity: Human and Horse. Does not react with mouse and rat. Others not tested.

Epitope: Not determined

Immunogen: Recombinant hMART-1 protein

Molecular Weight: 18kDa

Positive Control: CAC1 melanoma cells and melanomas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

MART-1/Melan-A (Melanoma Marker) Ab-1
Immunoprecipitation
Unconjugated
RUO
Primary
M2-7C10
Monoclonal
Murine
Cancer and Tumor Biology
Recombinant hMART-1 protein
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit