Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARVELD1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA520853

Catalog No. PIPA520853


Only null left
Add to Cart

Description

Description

A suggested positive control is mouse heart tissue lysate. PA5-20853 can be used with blocking peptide PEP-0967.

MARVELD1, (MARVEL domain-containing 1), is one of the MARVEL (MAL and related proteins for vesicle trafficking and membrane link) domain-containing proteins. It is a nuclear-localized protein that is widely expressed in normal human tissues. While it has been reported to be overexpressed in colon cancer, MARVELD1 protein levels are generally lower in several primary tumors including ovary, breast, kidney, bladder and liver. This lower level of MARVELD1 expression in tumors is thought to be due to a higher methylation status of the MARVELD1 promoter as treatment of MDA-MB-361 with 5-aza-2'-deoxycytidine restores MARVELD1 expression, suggesting that loss of MARVELD1 expression may contribute to tumorgenicity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MARVELD1
Polyclonal
Unconjugated
MARVELD1
AI504298; bA548K23.8; GB14; MARVD1; MARVEL (membrane-associating) domain containing 1; MARVEL domain containing 1; MARVEL domain-containing protein 1; Marveld1; MRVLDC1; putative MARVEL domain-containing protein 1; RP11-548K23.8
Rabbit
Antigen affinity chromatography
RUO
277010, 309375, 83742
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q7TQJ1, Q9BSK0
MARVELD1
A 16 amino acid peptide from near the amino terminus of human MARVELD1.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.