Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARVELD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30415020UL
Description
MARVELD1 Polyclonal specifically detects MARVELD1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MARVELD1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
83742 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
bA548K23.8, FLJ22343, FLJ23440, GB14, MARVD1, MARVEL (membrane-associating) domain containing 1, MARVEL domain containing 1, MGC4415, MRVLDC1, putative MARVEL domain-containing protein 1, RP11-548K23.8 | |
A synthetic peptide of Human MARVELD1. | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title