Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARVELD2/Tricellulin Rat anti-Mouse, Unconjugated, Clone: N54, MilliporeSigma™

Rat Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen MARVELD2/Tricellulin
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABT15341MI
View Documents
MilliporeSigma
MABT1534100UG
100 μg
Each for $372.00
Add to cart
 
MABT15342MI
View Documents
MilliporeSigma
MABT153425UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

MARVELD2/Tricellulin Monoclonal antibody specifically detects MARVELD2/Tricellulin in Mouse samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

MARVELD2/Tricellulin
Unconjugated
Rat
MARVEL domain-containing protein 2
GST-tagged recombinant fragment corresponding to 150 amino acids from the N-terminal region of murine Tricellulin.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Purified
Mouse
Marveld2, Mrvldc2, Tric
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit