Thermo Scientific™ MAS™ Cerebrospinal Fluid Control

Monitor the performance of quantitative and manual chemical analysis of human cerebrospinal fluid with Thermo Scientific™ MAS™ Cerebrospinal Fluid Control.

Manufacturer: Thermo Scientific™ 10014444

 View more versions of this product

Catalog No. 23-640-122

    N/A

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Description Cerebrospinal Fluid Control, Vista Vials, Assayed, Bi-level
    Analytes Albumin, Alpha-1-Globulin, Alpha-2-Globulin, Beta-Globulin, Chloride, Gamma-Globulin, Glucose, Glutamine, IgA, IgG, IgM, Lactic Acid, LDH, Oligoclonal Bands, Potassium, Prealbumin, Sodium, Total Protein,Albumin, Alpha-1-Globulin, Alpha-2-Globulin, Beta-Globulin, Chloride, Gamma-Globulin, Glucose, Glutamine, IgA, IgG, IgM, Lactic Acid, LDH, Oligoclonal Bands, Potassium, Prealbumin, Sodium, Total Protein
    Storage Requirements 24 month shelf life at 2° to 8°C; 7 days onboard instrument
    Packaging 6 Pk.
    Quantity 2mL,2mL
    Level Bi-level,Bi-level

    Monitor the performance of quantitative and manual chemical analysis of human cerebrospinal fluid with Thermo Scientific™ MAS™ Cerebrospinal Fluid Control, a bi-level control assayed for cellulose space acetate and agarose gel electrophoresis.

    • Requires no additional preparation
    • Eliminates variables due to reconstitution, mixing errors, or uncertain water quality
    • Contains 18 analytes† including Glutamine and Potassium
    • Human-serum based
    • Assayed ranges for both chemistry and protein electrophoresis
    • Assayed for most major analyzers
    • Refer to package insert for specific analyte and stability claims

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit