Thermo Scientific MAS™ DOA-XSE™ Controls
Monitor semi-quantitative drugs of abuse screening test methods with ready-to-use, liquid assayed multi-analyte controls.
Manufacturer: Thermo Scientific SDOA101
 View more versions of this product
Catalog No. 23100007
 N/A
N/A
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Description MAS DOA-XSE Controls
    Analytes Benzoylecgonine, Cannabinoid, d-Methamphetamine, Ethanol, LSD, Methadone, Methaqualone, Morphine, Oxazepam, Phencyclidine, Propoxyphene, Secobarbital,Benzoylecgonine, Cannabinoid, d-Methamphetamine, Ethanol, LSD, Methadone, Methaqualone, Morphine, Oxazepam, Phencyclidine, Propoxyphene, Secobarbital
    Storage Requirements 30 day open-vial stability for all analytes at 2° to 8°C
    Quantity 6 x 18mL,6 x 18mL
    Level Level 1 (Negative),Level 1 (Negative)
    Item Description Confirmation values provided by SAMHSA certified labs
    Monitor semi-quantitative drugs of abuse screening test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ DOA-XSE™ Controls. These controls are for use with a variety of clinical chemistry analyzers.
    The DOA-XSE contains SAMHSA 5, Barbiturates, Benzodiazepines, Ethanol, LSD Metadone, Methaqualone and Propoxyphene.
    • Monitor assay conditions in semi-quantitative and qualitative analyses of patient urine specimens for drugs and drug metabolites
    • Confirmation values provided by SAMHSA-certified labs
    • DOA-XSE for use with Syva™-Emit™ methods
    • Available in 4 distinctive levels
  • Description & Specifications
Related Products
Product Content Feedback