Learn More
Thermo Scientific MAS™ DOA-XSE™ Controls
Monitor semi-quantitative drugs of abuse screening test methods with ready-to-use, liquid assayed multi-analyte controls.
Manufacturer:  Thermo Scientific SDOA404
 View more versions of this product
Catalog No. 23-100010
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
Monitor semi-quantitative drugs of abuse screening test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ DOA-XSE™ Controls. These controls are for use with a variety of clinical chemistry analyzers.
The DOA-XSE contains SAMHSA 5, Barbiturates, Benzodiazepines, Ethanol, LSD Metadone, Methaqualone and Propoxyphene.
  • Monitor assay conditions in semi-quantitative and qualitative analyses of patient urine specimens for drugs and drug metabolites
  • Confirmation values provided by SAMHSA-certified labs
  • DOA-XSE for use with Syva™-Emit™ methods
  • Available in 4 distinctive levels
Description & Specifications
Specifications
MAS DOA-XSE Controls
Level 4 (High positive)
30 day open-vial stability for all analytes at 2° to 8°C
Multi-Analytes
Urine
Benzoylecgonine, Cannabinoid, d-Methamphetamine, Ethanol, LSD, Methadone, Methaqualone, Morphine, Oxazepam, Phencyclidine, Propoxyphene, Secobarbital
6 x 18mL
Control