Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAS™ UA Controls

Catalog No. p-4527736
Click to view available options
Description:
UA Controls, Assayed
Level:
1
2 (abnormal)
Quantity:
15 mL
60 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Monitor urine dipstick and microscopic components with bi-level controls with improved room temperature storage capability.

Monitor urine dipstick and microscopic components with Thermo Scientific™ MAS™ UA Controls, bi-level controls with improved room temperature storage capability. Liquid and ready-to-use, this urine control is assayed for most major reagents strips and strip systems. It includes 10 commonly-assayed dipstick tests plus Microalbumin, Creatinine and hCG.

  • Level 2 (abnormal)
  • Available in convenient 15mL dropper bottles
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description UA Controls, Assayed
Detectable Analytes Bilirubin, Blood, Creatinine, Crystals, Glucose, hCG, Ketones, Leukocyte Esterase, Microalbumin, Nitrite, Osmolality, pH, Potassium, Protein, Red Blood Cells, Sodium, Specific Gravity, Urobilinogen, White Blood Cells
Sample Type Urine
Type Control
Level 2 (abnormal)
Packaging 6 Pk.
Quantity 15 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.