Learn More
Thermo Scientific™ MAS™ UrichemTRAK Controls, Vista Vials, 6 x 2mL
Monitor the most commonly tested urine chemistry analytes with liquid, assayed Thermo Scientific™ MAS™ UrichemTRAK Controls, available in a convenient vial format
Supplier:  Thermo Scientific™ 10014127
Catalog No. 23-139-630
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Monitor the most commonly tested urine chemistry analytes with liquid, assayed Thermo Scientific™ MAS™ UrichemTRAK Controls, available in a convenient Vista™ vial format. Ready-to-use and requiring no additional preparation, these controls will not interfere with ISEs.
  • Liquid, human urine matrix
  • Open and use; no equilibration required
  • Two-year open-vial stability
  • Individual levels or multi-pack configurations
  • Stable for 24 months at 2° to 8°C
  • Assayed values available for most major analyzers