Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MASH1 Mouse anti-Mouse, Rat, Clone: 24B72D11, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 14579480

 View more versions of this product

Catalog No. 50-112-2693


Add to cart

Description

Description

Description: The monoclonal antibody 24B72D11 recognizes mouse and rat Mammalian achaete-scute homolog 1 (MASH1), a 34 kDa basic helix-loop-helix (bHLH) transcription factor. MASH1, like family members Olig2 and Ngn2, is expressed in progenitor cells throughout the nervous system and retina and is a key regulator of cell fate specification. Expression of MASH1 is regulated by the Delta/Notch1 pathway through direct binding of Hes1 to MASH1 as well as competitive binding to MASH1 hetero-dimerizing proteins. At a transcriptional level, MASH1 expression is controlled by notch-mediated regulation of sox2, which binds to the MASH1 promoter region up-regulating expression. MASH1 has been implicated in the formation of multiple neural cell types including dopaminergic, serotonergic, glutaminergic and GABAergic neurons. Overexpression of MASH1 has been shown in small cell lung carcinoma and medullary thyroid cancer. Applications Reported: This 24B72D11 antibody has been reported for use in western blotting, immunohistochemical staining of frozen tissue sections, and immunocytochemistry. Applications Tested: This 24B72D11 antibody has been tested by western blot on reduced PC12 cells and can be used at less than or equal to 5 µg/mL. The 24B72D11 antibody has also been tested by immunocytochemistry on fixed and permeablized PC12 cells at can be used at less than or equal to 20 µg/mL.

It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered. ASCL1 (also known as ASH1) is a basic helix-loop-helix transcription factor that is required for early development of the nervous system. Expressed in fetal brain, ASCL1 is essential for the proper development of autonomic neurons and for the survival of subsets of autonomic neurons. ASCL1 interaction with MEF-2A may regulate the expression of specific genes that are critical for the formation of distinct neuronal circuits within the central nervous system. The high level of ASCL1 expression in neuroendocrine tumors, such as medullary thyroid cancer, small cell lung cancer and lung cancer with neuroendocrine features may provide a useful marker for cancers with neuroendocrine features. Mapping to human chromosome 12, the ASCL1 gene contains a trinucleotide repeat region, making this locus a candidate for inherited disease.
Specifications

Specifications

MASH1
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q02067, P19359
KMT2H
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
24B72D11
Unconjugated
Ascl1
Liquid
Ascl1
IgG1, kappa
25 μg
4° C
Primary
17172, 64186
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.