Learn More
Selleck Chemical LLC
Masitinib (AB1010) 10mg 790299-79-5
Manufacturer: Selleck Chemical LLC S106410MG
Masitinib is a novel inhibitor for Kit (c-Kit) and PDGFRα/β with IC50 of 200 nM and 540 nM/800 nM, weak inhibition to ABL and c-Fms. Phase 3.