Learn More
Selleck Chemical LLCSupplier Diversity Partner Masitinib (AB1010) 200mg 790299-79-5  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S1064200MG
Masitinib is a novel inhibitor for Kit (c-Kit) and PDGFRα/β with IC50 of 200 nM and 540 nM/800 nM, weak inhibition to ABL and c-Fms. Phase 3.
Catalog No. 50-649-8