Learn More
Thermo Scientific™ Mass Spectrometry Sample Preparation Bundle
Mass Spectrometry Sample Preparation Bundle combines high quality validated reagents to enable success in sample preparation for MS-based analysis. This bundle includes Pierce Tryspin and Lys-C Proteases, Pierce Peptide Desalting Spin Columns and Pierce Quantitative Colorimetric Peptide Assay kit
Manufacturer:  Thermo Scientific™ B90012
Catalog No. PIB90012
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Pierce Trypsin Protease MS Grade (5 x 20 µg), Pierce Lys-C Protease MS Grade (100 µg), Pierce Peptide Desalting Spin Columns (25 columns), Pierce Quantitative Colorimetric Peptide Assay (500 assays)
Description
Description
  • Thermo Scientific Pierce Trypsin Protease, MS Grade, and Pierce Lys-C Protease, MS Grade are highly purified proteases that have been validated for maximum activity and stability in proteomic applications
  • Thermo Scientific Pierce Peptide Desalting Spin Columns are ready-to-use centrifuge spin columns that enable efficient desalting of peptide samples following enzymatic digestion
  • The Thermo Scientific Pierce Quantitative Colorimetric Peptide Assay is an easy-to-use colorimetric microplate assay designed specifically to improve the sensitivity and reproducibility of quantitation of peptide mixtures
Specifications
Specifications
Pierce Trypsin Protease MS Grade (5 x 20 µg), Pierce Lys-C Protease MS Grade (100 µg), Pierce Peptide Desalting Spin Columns (25 columns), Pierce Quantitative Colorimetric Peptide Assay (500 assays)
1 Bundle
See instructions for individual components