Learn More
Alfa Aesar™ Mast Cell Degranulating Peptide HR-1
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J63655LB0
 View more versions of this product
Catalog No. AAJ63655LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1492.96
5mg
Powder
RUO
HPLC
Degranulates mast cells, initiating histamine release
Ile-Asn-Leu-Lys-Ala-Ile-Ala-Ala-Leu-Val-Lys-Lys-Val-Leu-NH2
Mast Cell Degranulating Peptide HR-1