Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Mast Cell Tryptase Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ Mast Cell Tryptase Ab-2, Mouse Monoclonal Antibody.

$456.00 - $972.00

Specifications

Antigen Mast Cell Tryptase Ab-2
Clone AA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1216P1
View Documents
Thermo Scientific™
MS1216P1
Purified Ab with BSA and Azide 500μL
Each for $456.00
Add to cart
 
MS1216P1ABX
View Documents
Thermo Scientific™
MS1216P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $501.50
Add to cart
 
MS1216PABX
View Documents
Thermo Scientific™
MS1216PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $972.00
Add to cart
 
MS1216R7
View Documents
Thermo Scientific™
MS1216R7
Ready-to-use for IHC (UVLP) 7mL
Each for $570.50
Add to cart
 
MS1216R1
View Documents
Thermo Scientific™
MS1216R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
MS1216P0
View Documents
Thermo Scientific™
N
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS1216P
View Documents
Thermo Scientific™
MS1216P
Purified Ab with BSA and Azide 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ Mast Cell Tryptase Ab-2, Mouse Monoclonal Antibody.

Mast cells contain a number of preformed chemical mediators such as histamine, chymase, carboxypeptidase and proteolytic tryptase. A substantial quantity of tryptase is estimated to be found in mast cells of skin and lung and suggestes this enzyme plays a major role in mast cell mediated events. In vitro studies indicate tryptase can cleave C3 to form C3a anaphylatoxin, inactivate fibrinogen as a coaguable substrate for thrombin and activate latent collagenase. Human mast cell tryptase has considerable potential as a marker for mast cell activation as well as being a mediator of inflammation.

Host Species: Mouse

Clone: AA1

Isotype: IgG1

Species Reactivity: Human, Monkey, Dog and Cat. Others not known.

Epitope: Not determined

Immunogen: Human mast cell tryptase purified from human lung tissues

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Mast Cell Tryptase Ab-2
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RUO
Primary
IgG1
AA1
Monoclonal
Murine
Enzyme
Human mast cell tryptase purified from human lung tissues
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit