Learn More
Cole-Parmer Masterflex L/S Digital Drive, 600 rpm; 115/230 VAC  DFS Item
Manufacturer:  Cole-Parmer 0752220
These precision drives offer analog remote control options and simple programming—ideal for automated process applications. Drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Motor is reversible so you can purge tubing before or after pumping and pump fluid in either direction.
Catalog No. NC1089752