Learn More
Master Lock™ Laminated Steel Padlock
Manufacturer:  Master Lock 7KAD
 View more versions of this product
Catalog No. 19-133-770
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.18 in.
Gray bumper
Keyed alike
Industrial, storefront and business gates, tool cribs, construction sites and equipment, vending machines
0.56 in.
29mm
Dual locking lever
0.51 in.
13mm
5mm
Master lock padlock
Steel
1.12 in.
14mm
4-pin W7 cylinder, color bumper, dual locking lever
1.12 in.
29mm