Learn More
Master Lock™ Safety High-Visibility Aluminum Padlock
Manufacturer:  Master Lock™ 411ORJ
 View more versions of this product
Catalog No. 19-813-522
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Withstands harsh, corrosive environments; available in 6 vibrant colors; Over 5-pin cylinder with over 10,000 key changes
  • Durable padlock protection with a high-visibility, powder-coated finish
  • Vibrant colors provide safety color recognition plus high security
  • These aluminum locks have a pick-resistant five-pin Pro Series™ cylinder, but are conveniently rekeyable Corrosive-resistant locks are all 1 1/2 in. wide (40mm) and have shackle diameters of 1/4 in. (6.35mm)
  • Vertical shackle clearance is 1 1/16 in. (27mm)
  • Uses FDA approved lubricants safe for use in food processing