Learn More
Masterflex™ L/S™ Analog Variable-Speed Console Drives
Masterflex™ L/S™ Analog Variable-Speed Console Drives are compact and cost effective for general fluid transfer applications
 View more versions of this product
Catalog No. 13-200-022
Add to cart
Description
Description
  • Flow rate 0.42 to 2900 mL/min with L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Reversible motor lets you easily prime/purge tubing and pump in either direction
  • Remote control of start/stop via DB9 connector on back of drive
  • Specifications
    Specifications
    180 to 260 VAC
    Analog
    1.2 to 2900 mL/min
    6 ft. (1.8 m) line cord with IEC 320/CEE22 socket. Shipped with country-specific cord/plug set; Please specify ultimate destination when ordering.
    Yes
    20 to 600 rpm
    50/60 Hz
    1 pump head
    Painted Steel and ABS Plastic