Learn More
Cole-Parmer L/S economy variable-speed drive, 20 to 600 rpm, 115 VAC.  DFS Item
Manufacturer:  Cole-Parmer 0755500
Masterflex 07555-00 L/S Variable-Speed Console Drive, 20 to 600 rpm, 115 VAC, Compact and cost effective for general fluid transfer applicationsFlow range: 0.42 to 2900 mL/minSimple operation with single-turn speed controlSeparate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power onReversible motor lets you easily prime/purge tubing and pump in either directionRemote control of start/stop via DB9 connector on back
Catalog No. NC1627944