Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Masterflex™ L/S™ Precision Pump Tubing, Norprene™ Food A-60-F

Masterflex™ L/S™ Precision Pump Tubing has FDA approval for food and beverage applications

$80.80 - $161.60

Specifications

Autoclavable Yes
Color Tan
For Use With (Application) Food, dairy, and beverage industries
Material Norprene Food
Overall Length (English) 50 ft.
View More Specs

Products 4
Catalog Number Tubing Size Price Quantity & Availability  
Catalog Number Tubing Size Price Quantity & Availability  
13-200-027
View Documents
L/S 13
Pack for $80.80
Add to cart
 
13-200-028
View Documents
L/S 14
Pack for $84.10
Add to cart
 
13-200-029
View Documents
L/S 18
Pack for $161.60
Add to cart
 
13-200-031
View Documents
L/S 25
Pack for $119.50
Add to cart
 
Description

Description

  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • Applications: Ideal for the food, dairy, and beverage industries
  • Characteristics: Heat, ozone, and UV light resistant, non-aging, non-oxidizing, superior acid and alkali resistance, opaque beige
Specifications

Specifications

Yes
Food, dairy, and beverage industries
50 ft.
Tan
Norprene Food
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit