Learn More
Cole-Parmer L/S Computer-Compatible Digital Drive, 0.02 to 100 rpm, 115/230 VAC
Manufacturer:  Cole-Parmer 0755130
Encompass_Preferred
Masterflex L/S Computer-Compatible Digital Drive, 0.02 to 100 rpm, 115/230 VAC, Interface these digital drives to a PC or network for automated lab and process applications--or use as a stand-alone dispensing pumpFlow rate 0.001 to 3400 mL/min with L/S tubing. Flow rate depends on drive rpm and tubing size.Graphical LCD shows four operating modes continuous run, timed dispense, copy dispense, volume dispense. View pump performance continually.Interface with a PC via RS-232 or USB connection Dispense by Volume in mL (0.001 to 99999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval, Type of Pump Heads Accepted- Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm
Catalog No. NC0927195
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.