Learn More
Cole-Parmer Masterflex L/S Digital Precision Modular Drive with Remote I/O and Benchtop Controller, 1 to 100 rpm; 90 to 260 VAC
Manufacturer:  Cole-Parmer 0755710
Encompass_Preferred
L/S modular drives feature reversible pumping and allow you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering applications.
Catalog No. NC0458484
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.