Learn More
Cole-Parmer Masterflex I/P Precision Pump Tubing, BioPharm Platinum-Cured Silicone, I/P 26; 25 ft  DFS Item
Manufacturer:  Cole-Parmer 9642026
Choose our BioPharm Silicone platinum-cured tubing for purity, very low extractables, and an ultra-smooth inner surface. Ideal for pharmaceutical and biotechnological applications, this tubing complies with USP class VI and 3A sanitary standards. The ultra-smooth inner surface minimizes particle entrapment and microscopic build-up during fluid transfer. Tubing can be sterilized by autoclave, gamma radiation, or ethylene oxide.
Catalog No. 50-677-58