Learn More
Alfa Aesar™ Mastoparan
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J61662MCR
 View more versions of this product
Catalog No. AAJ61662MCR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
1478.93
1mg
Powder
RUO
Mastoparan
HPLC
A peptide toxin originally isolated from wasp venom; stimulates exocytosis
Ile-Asn-Leu-Lys-Ala-Leu-Ala-Ala-Leu-Ala-Lys-Lys-Ile-Leu-NH2
-30° to -10°C