Learn More
Invitrogen™ MAT1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117063
Description
Positive Samples: HepG2, 293T, Mouse liver, Rat liver Immunogen sequence: MNGPVDGLCD HSLSEGVFMF TSESVGEGHP DKICDQISDA VLDAHLKQDP NAKVACETVC KTGMVLLCGE ITSMAMVDYQ RVVRDTIKHI GYDDSAKGFD FKTCNVLVAL EQQSPDIAQC VHLDRNEEDV GAGDQGLMFG YATDETEECM PLTIILAHKL NARMADLRRS GLLPWLRPDS KTQVTVQYMQ DNGAVIPVRI HTIVISVQHN EDITLEEMRR ALKEQVIRAV VPAKYLDEDT VYHLQPSGRF VIGGPQGDAG VTGRKIIVDT YGGWGAHGGG AFSGKDYTKV DRSAAYAARW VAKSLVKAGL CRRVLVQVSY AIGVAEPLSI SIFTYGTSQK TERELLDVVH KNFDLRPGVI VRDLDLKKPI YQKTACYGHF GRSEFPWEVP RKLVF.
MAT1A catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and dominant mutations, the latter identified in autosomal dominant persistant hypermethioninemia.This gne encodes methionine adenosyltransferase I (alpha isoform), which catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and dominant mutations, the latter identified in autosomal dominant persistant hypermethioninemia.
Specifications
MAT1A | |
Polyclonal | |
Unconjugated | |
MAT1A | |
AdoMet; AdoMet synthase 1; adoMet synthetase 1; AI046368; Ams; Ams1; Cartilage matrix protein; CMP; CRTM; MAT; MAT 1; Mat1a; MATA1; MAT-I/III; MATN1; matrilin 1, cartilage matrix protein; Matrilin-1; Methionine adenosyltransferase 1; methionine adenosyltransferase 1A; methionine adenosyltransferase I, alpha; methionine adenosyltransferase I/III; S - adenosylmethionine synthetase; SADE; S-adenosylmethionine synthase; S-adenosylmethionine synthase isoform type-1; S-adenosylmethionine synthase isoform type-1; S-adenosylmethionine synthase; S-adenosylmethionine synthetase isoform type-1; SAMS; SAMS1; SAS; similar to S-adenosylmethionine synthetase alpha and beta forms (Methionine adenosyltransferase) (AdoMet synthetase) (MAT-I/III); wu:fi35e01; zgc:55442 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11720, 25331, 4143 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.8 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P13444, Q00266, Q91X83 | |
MAT1A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-395 of human MAT1A (NP_0004201). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.