Learn More
Invitrogen™ MAT2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588170
Description
Immunogen sequence: MVGREKELSI HFVPGSCRLV EEEVNIPNRR VLVTGATGLL GRAVHKEFQQ NNWHAVGCGF RRARPKFEQV NLLDSNAVHH IIHDFQPHVI VHCAAERRPD VVENQPDAAS QLNVDASGNL AKEAAAVGAF LIYISSDYVF DGTNPPYREE DIPAPLNLYG KTKLDGEKAV LENNLGAAVL RIPILYGEVE KLEESAVTVM FDKVQFSNKS ANMDHWQQRF PTHVKDVATV CRQLAEKRML DPSIKGTFHW SGNEQMTKYE MACAIADAFN LPSSHLRPIT DSPVLGAQRP RNAQLDCSKL ETLGIGQRTP FRIGIKESLW PFLIDKRWRQ TVFH; Positive Samples: HeLa, Mouse brain, Mouse kidney.
MAT2B belongs to the methionine adenosyltransferase (MAT) family. MAT catalyzes the biosynthesis of S-adenosylmethionine from methionine and ATP. This protein is the regulatory beta subunit of MAT. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Specifications
MAT2B | |
Polyclonal | |
Unconjugated | |
MAT2B | |
1110064C04Rik; 2410018D16Rik; AI182287; AU022853; beta regulatory subunit of methionine adenosyltransferase; dTDP-4-keto-6-deoxy-D-glucose 4-reductase; MAT II beta; MAT2B; MAT-II; MATIIbeta; methionine adenosyltransferase 2 beta subunit; methionine adenosyltransferase 2 subunit beta; methionine adenosyltransferase 2B; Methionine adenosyltransferase II beta; methionine adenosyltransferase II, beta; MSTP045; Nbla02999; Putative dTDP-4-keto-6-deoxy-D-glucose 4-reductase; putative protein product of Nbla02999; SDR23E1; short chain dehydrogenase/reductase family 23E, member 1; testicular tissue protein Li 118; TGR; UNQ2435/PRO4995 | |
Rabbit | |
Affinity chromatography | |
RUO | |
108645, 27430 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.76 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q99LB6, Q9NZL9 | |
MAT2B | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-334 of human MAT2B (NP_0374151). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.