Learn More
Sklar InstrumentsSupplier Diversity Partner TC Mathieu Needle Holder, Sklar, 5-1/2 in., Blunt, Curved, Cross-Serrated, Pliers, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 218073
TC Mathieu Needle Holder, Sklar, 5-1/2 in., Blunt, Curved, Cross-Serrated, Pliers, Tungsten Carbide / Stainless Steel. Sklar's Mathieu TC Needle Holder is a single spring instrument with a plier handle grip that curves inward at the bottom and features a ratchet locking mechanism at the base of the handles that can be easily squeezed to be locked and unlocked. This needle holder is used when more refined suturing is required. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles. This product is curved with cross-serrated jaws, and an overall length of 5-1/2 inches.
Catalog No. 50-143-7758