Learn More
Sklar InstrumentsSupplier Diversity Partner Mathieu Delicate Needle Holder, Sklar, 5-1/2 in., Blunt, Straight, Cross-Serrated, Pliers with Lock, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 218105
Mathieu Delicate Needle Holder, Sklar, 5-1/2 in., Blunt, Straight, Cross-Serrated, Pliers with Lock, Stainless Steel. Sklar's Mathieu Needle Holder is a single spring instrument with a plier handle grip that curves inward at the bottom and features a ratchet locking mechanism at the base of the handles that can be easily squeezed to be locked and unlocked. This needle holder is used when more refined suturing is required. This product is straight with delicate cross-serrated jaws, and an overall length of 5-1/2 inches.
Catalog No. 50-143-7786