Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Matrigel™ Growth Factor Reduced (GFR) Basement Membrane Matrix, LDEV-Free
Click to view available options
Quantity:
100 mL (10 mL x 10)
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Specifications
Specifications
Content And Storage | Store at -20°C. Avoid multiple freeze-thaws. Do not store in frost-free freezer. Keep frozen. |
Without Additives | No LDEV |
For Use With (Application) | Cell Growth and Differentiation, Metabolism/Toxicology Studies, Invasion Assays, In Vitro and In Vivo Angiogenesis Assays, In Vivo Angiogenesis Studies and Augmentation of Tumors in immunosuppressed Mice |
Quantity | 100 mL (10 mL x 10) |
Source | Engelbreth-Holm-Swarm (EHS) Mouse Tumor |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction