Learn More
Corning™ Matrigel™ Growth Factor Reduced (GFR) Basement Membrane Matrix, LDEV-Free
Suited for applications requiring highly defined basement membrane preparation.
Specifications
Content And Storage | Store at -20°C. Avoid multiple freeze-thaws. Do not store in frost-free freezer. Keep frozen. |
---|---|
Without Additives | No LDEV |
For Use With (Application) | Cell Growth and Differentiation, Metabolism/Toxicology Studies, Invasion Assays, In Vitro and In Vivo Angiogenesis Assays, In Vivo Angiogenesis Studies and Augmentation of Tumors in immunosuppressed Mice |
Source | Engelbreth-Holm-Swarm (EHS) Mouse Tumor |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
CB356252
|
Corning™
356252 |
50 mL (10 mL x 5) | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
CB356253
|
Corning™
356253 |
100 mL (10 mL x 10) | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Specifications
Store at -20°C. Avoid multiple freeze-thaws. Do not store in frost-free freezer. Keep frozen. | |
No LDEV | |
Cell Growth and Differentiation, Metabolism/Toxicology Studies, Invasion Assays, In Vitro and In Vivo Angiogenesis Assays, In Vivo Angiogenesis Studies and Augmentation of Tumors in immunosuppressed Mice | |
Engelbreth-Holm-Swarm (EHS) Mouse Tumor |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.