Learn More
Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips), Extended Length
Achieve ultimate performance in automated pipetting with Thermo Scientific™ D.A.R.T.'s™.
Manufacturer:  Thermo Scientific™ 5417
 View more versions of this product
Catalog No. 14-387-689
Add to cart
Description
Description
Achieve ultimate performance in automated pipetting with Thermo Scientific™ D.A.R.T.'s™. Unlike conventional automation tips, D.A.R.T.'s are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision. Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x2, Matrix™ PlateMate™ 2x3, PlateMate™ Plus Systems and Versette.
Features:
  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access.
  • Highly polished molds and cores ensure low liquid retention.
  • Small internal tip orifice provides better small volume droplet formation
  • Sterile, sterile filtered, wide-bore, and extended-length tip options are available
  • Tips are thoroughly inspected for proper formation and straightness
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing.
  • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination.
Includes: 20 magazines per case
Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x3 and PlateMate™ Plus Systems.
Specifications
Specifications
20 x 384 tips
30μL
Sterile
Thermo Scientific™ Matrix Hydra DT, Thermo Scientific™ PlateMate Plus, Thermo Scientific™ Versette
384/100 Versette Pipetting Head
Non-Filtered
Extended Length
D.A.R.T.'S, Matrix
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.