Thermo Scientific™ Matrix™ Equalizer Electronic Multichannel Pipettes
This product is no longer being manufactured, although we will continue to provide spare parts until December 2020.
Manufacturer: Thermo Scientific™ 2131
 View more versions of this product
Catalog No. 14-387-135
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Accuracy ±2%
    Description Discontinued; 8 channel, 2 to 125µL
    Increments 0.1µL
    Precision ±1.0
    Color Yellow,Yellow
    Number of Channels 8
    Volume (Metric) Range 2 to 125µL
    Channels 8
    We will continue to offer Thermo Scientific™ Matrix pipette tips for your pipettes, however this product is no longer being manufactured, although we will continue to provide spare parts until December 2020. After this date it may be difficult to source parts. Please contact your sales representative for availability.
    The NEW Thermo Scientific™ E1-ClipTip™ Electronic Pipettes are replacing this product line. Please take a moment to learn more about sealed in security.
    If you require additional information, please contact your sales representative or our service department for assistance. You can still find a full listing of Matrix Pipette Tips.
  • Description & Specifications
Related Products
Product Content Feedback