Thermo Scientific™ Matrix™ Filtered Pipette Tips Available on GSA/VA Contract for Federal Government customers only.
Eliminate pipetter barrel contamination with Thermo Scientific™ Matrix™ Filtered Pipette Tips.
Manufacturer: Thermo Scientific™ 7275
 View more versions of this product
Catalog No. 14-387-173
  Check Availability
Add to Cart
    Specifications
    Packaging 10 x 96/Rack
    For Use With (Equipment) Gilson/Rainin P200
    Volume (Metric) 200μL
    Product Line Matrix
    Filtered Yes
    Sterility Sterile
    Model Thermo Scientific Matrix Electronic Pipetters
    Pipette Brand All, Matrix Impact 125¦L and 250¦L, Gilson/Rainin P200
    Eliminate pipetter barrel contamination with Thermo Scientific™ Matrix™ Filtered Pipette Tips. Rigorously tested to ensure optimum performance, these filtered tips eliminate the risk of cross-contamination from aerosols created while pipetting genomic material, bacterial samples or other precious samples.
    Benefits:
    • Stringent testing guarantees filter tips that effectively block aerosols.
    • Hydrophobic polyethylene filter material ensures no particulates contaminate samples.
    • Inadvertent contact with filter will not contaminate sample or result in sample loss.
    • Easy to open packaging maintains product sterility while providing easy access.
    • Hinged lid allows rack to be opened without laying lid on benchtop, helping maintain sterility.
    • Racks are sealed in thick polyethylene bag for sterility, which is easy to open with simple snap-and-tear packaging.
    Stringent Filter Testing:
    • Filter must meet certain porosity requirements and must form a complete barrier across face of tip.
    • To ensure defects are not present, two specific tests are performed: mercury intrusion and particle count testing.
    • Mercury Intrusion Testing (QC 13.1 - QC 13.8) ensures the filter itself meets specific aerosol blocking requirements.
    • Particle Count Testing (Test Protocol QAS-0100) ensures filter is effective at blocking aerosols after installation into tip; also ensures proper shape and fit of filter.
    • Every lot of tips is tested by drawing a vacuum of aerosols against the filter and ensuring particles do not pass through.
    Recommended for:
    • Bacteriology
    • Gel loading
    • PCR setup
    • Preventing cross-contamination during routine pipetting
  • Description & Specifications
Related Products
Product Content Feedback