Thermo Scientific™ Matrix™ Filtered TallTip Extended-Length Pipette Tips Available on GSA/VA Contract for Federal Government customers only.

Protect your samples and pipettes from contamination with filtered, extended-length narrow-tip pipette tips that maximize reach and minimize risk.

Manufacturer: Thermo Scientific™ 7635

 View more versions of this product

Catalog No. 14-387-149

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Packaging 10 x 96/Rack
    For Use With (Equipment) Universal Fit
    Volume (Metric) 12.5μL
    Product Line Matrix TallTip
    Filtered Yes
    Sterility Nonsterile
    Volume Range (Metric) 12.5µL

    Protect your samples and pipettes from contamination with Thermo Scientific™ Matrix™ Filtered TallTip Extended-Length Pipette Tips, which allow access to the bottom of test tubes, reagent bottles, flasks and other vessels without touching the barrel of your pipetter against the side of the tube. Non-beveled, narrow-tip ends provide cleaner touch-off of sidewalls for greater precision. The hydrophobic filters effectively block aerosols and ensure that no particulates contaminate samples.

    Features:

    • Prevent cross-contamination from vessel walls
    • Allow the use of large volume tips in small spaces, resulting in less liquid displacement
    • Reach the bottom of particularly long or narrow vessels that standard tips cannot reach
    • 102mm maximum length for safe access to 100mm test tubes
    • Flo-Thru rack taht reduces tip wobble and improves airflow while autoclaving
    • Universal fit that works with most major brands of pipetters

    Hydrophobic Filter:

    • Stringent testing guarantees filter tips that effectively blocks aerosols.
    • Hydrophobic filter material ensures no particulates contaminate samples.
    • Easy-to-open packaging ensures product maintains sterility while being easy to access.
    • Hinged rack lids can be opened without laying lid on benchtop, helping maintain sterility.

    Matrix Pipet Tip Features:

    • Sealing rings on tips ensure a confident seal without banging tips on a pipetter.
    • Highly polished molds and rigorous maintenance procedures ensure low liquid retention tips without special treatments or additives.
    • Stabilization rings in racks hold tips steady when seated on a pipetter.
    • Rigid rack design ensures solid, even tip seating.
    • Venting features in rack ensure proper heat flow during autoclaving and reduce condensation on tips.
    • Hinged lid prevents sterile lid from coming in contact with lab bench.
    • Finger notch on racks allows easy removal/insert of refill trays.
    • Snap-and-tear bags make it easy to open each individually wrapped sterile rack.
    • Well labeled racks simplify record keeping, tip identification and re-ordering.
    • Environmentally sensitive reduced rack size reduces waste.
    • Racks utilize recycled material and are marked with appropriate symbols to support recycling initiatives.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit