Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Modifier, Waste Water - Simulated, MilliporeSigma™ Supelco™

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

$178.60 - $179.70

Specifications

CAS 10035-04-8
Grade Certified Reference Material
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-100-2622
View Documents
MilliporeSigma Supelco
MMW001250ML
250 mL
Each for $179.70
Add to cart
 
11-100-2623
View Documents
MilliporeSigma Supelco
MMW0015X20ML
5 x 20 mL
Each for $178.60
Add to cart
 
Description

Description

Featured Industries: Environmental
Specifications

Specifications

10035-04-8
Certified Reference Material
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit