Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Matrix™ 384 Format Pipette Tips

Choose from a comprehensive selection of pipette tips.

$654.00 - $702.00

Specifications

For Use With (Equipment) Thermo Scientific™ Matrix Pipettor
Product Line Matrix
Filtered Non-filtered
View More Specs

Products 4
Catalog Number Mfr. No. Volume (Metric) Sterility Format Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Sterility Format Price Quantity & Availability  
14-387-191
View Documents
Thermo Scientific™
7421
12.5 μL Non-sterile Rack Case of 3840 for $654.00
Add to cart
 
14-387-192
View Documents
Thermo Scientific™
7422
12.5 μL Sterile Rack Case of 3840 for $702.00
Add to cart
 
14-387-194
View Documents
Thermo Scientific™
7431
30 μL Non-sterile Rack Case of 3840 for $654.00
Add to cart
 
14-387-195
View Documents
Thermo Scientific™
7432
30 μL Sterile Rack Case of 3840 for $702.00
Add to cart
 
Description

Description

Choose from a comprehensive selection of pipette tips. Thermo Scientific™ Matrix 384 Format Pipette Tips are supplied in innovative new Thermo Scientific™ Flo-Thru™ racks. Matrix pipetters and pipet tips are designed together to ensure that a perfect balance exists between security of the seal and ease of application/ejection. Manufactured under tight QC specifications, these standard tips rival special low-retention tips currently on the market after pipetting fluids such as dH2O, 50% Glycerol, Hybridized Buffer and 0.1% SDS.

Benefits:

  • Sealing rings on tips ensure confident, leak-free seal without banging tips on pipetter.
  • Because the sealing rings compress, these tips also offer a secure seal on most other pipetters on the market.
  • Highly polished molds and rigorous maintenance procedures ensure low liquid retention tips without special treatments or additives.
  • Stabilization rings in racks hold tips steady when seating on pipetter.
  • Venting features in rack ensure proper heat flow during autoclaving and reduce condensation on tips.
  • Finger notch on racks allows easy removal/insertion of refill trays.

Innovative Racks and Packaging:

  • Rigid rack design ensures solid, even tip seating; rack will not flex or bend when trying to apply tips.
  • Unique Thermo Scientific™ Flo-Thru™ rack individually supports tips to prevent them from wobbling when trying to seat them on the pipetter.
  • Hinge prevents sterile lid from coming in contact with lab bench.
  • Venting along the edge or around each tip improves heat flow during autoclaving; reduces condensation that can form on the tips.
  • Dual-position lid can be propped open to prevent vacuum deformation during autoclaving.
  • Sterile racks are sold in individual bags with quick-tear openings; each rack remains sterile until ready for use.
  • Each rack is labeled with product details including re-ordering information, description, volume range, and lot number.
  • Racks are constructed of clean recycled materials and are made as small as possible to reduce waste.
  • Racks are marked with appropriate symbols to support recycling initiativ
Specifications

Specifications

Thermo Scientific™ Matrix Pipettor
Matrix
Non-filtered
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit