Learn More
Thermo Scientific™ Matrix™ Pipette Tips In Ecotips Refill Packaging System
Process samples quickly with fast rack reloading in an environmentally friendly, space-saving design with this unique tip refill system.
Manufacturer:  Thermo Scientific™ 8246
 View more versions of this product
Catalog No. 14-387-162
  Check Availability
Add to cart
Specifications
Description
Process samples quickly with fast rack reloading in an environmentally friendly, space-saving design with the Thermo Scientific™ Matrix™ Pipette Tips In Ecotips Refill Packaging System, which reduces plastic waste by eliminating the need for a new tip box for each rack of tips. This system is 100% recyclable and each component is marked with appropriate recycling symbols. Trays of tips are nested in easily accessible packaging that takes up very little space and eliminates the need to dispose of a tip box each time a rack is emptied.
  • Compact alternative to standard racked tips
  • Quick, efficient refill; reloading empty tip boxes is quick and easy
  • Made from 100% virgin polypropylene; insert trays and boxes come from our own recycled materials
  • Saves bench, shelf and closet space; refills fit easily in a drawer, on a shelf or on the bench.
  • Allows for quick and efficient transfer of tips from packaging to empty tip box
  • Finger tabs on tip trays for easy removal of empty inserts from each tip box
  • Eliminates the need for mechanical towers, clumsy stacks of tips or other transfer mechanisms
Description & Specifications
Specifications
TallTip ECOTIPS
No
Reload
Thermo Scientific Matrix Pipettes
1250μL
Nonsterile
Refill Trays, 10 x 96
Ecotips