Learn More
Thermo Scientific Matrix Pipette Tips
Choose from a comprehensive selection of pipette tips.
Manufacturer: Thermo Scientific 732111
Description
Choose from a comprehensive selection of pipette tips. Thermo Scientific Matrix Pipette Tips are supplied in innovative new Thermo Scientific Flo-Thru racks. Matrix pipetters and pipet tips are designed together to ensure that a perfect balance exists between security of the seal and ease of application/ejection. Manufactured under tight QC specifications, these standard tips rival special low-retention tips currently on the market after pipetting fluids such as dH2O, 50% Glycerol, Hybridized Buffer and 0.1% SDS.
Benefits:
- Sealing rings on tips ensure confident, leak-free seal without banging tips on pipetter.
- Because the sealing rings compress, these tips also offer a secure seal on most other pipetters on the market.
- Highly polished molds and rigorous maintenance procedures ensure low liquid retention tips without special treatments or additives.
- Stabilization rings in racks hold tips steady when seating on pipetter.
- Venting features in rack ensure proper heat flow during autoclaving and reduce condensation on tips.
- Finger notch on racks allows easy removal/insertion of refill trays.
Innovative Racks and Packaging:
- Rigid rack design ensures solid, even tip seating; rack will not flex or bend when trying to apply tips.
- Unique Thermo Scientific Flo-Thru rack individually supports tips to prevent them from wobbling when trying to seat them on the pipetter.
- Hinge prevents sterile lid from coming in contact with lab bench.
- Venting along the edge or around each tip improves heat flow during autoclaving; reduces condensation that can form on the tips.
- Dual-position lid can be propped open to prevent vacuum deformation during autoclaving.
- Sterile racks are sold in individual bags with quick-tear openings; each rack remains sterile until ready for use.
- Each rack is labeled with product details including re-ordering information, description, volume range, and lot number.
- Racks are constructed of clean recycled materials and are made as small as possible to reduce waste.
- Racks are marked with appropriate symbols to support recycling initiatives.
Specifications
Thermo Scientific™ Matrix Pipettor | |
300μL | |
Non-sterile | |
Matrix |
Rack | |
Non-Filtered | |
Thermo Scientific™ Matrix Pipettor |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.