Learn More
Thermo Scientific™ Matrix™ Pipette Tips
Choose from a comprehensive selection of pipette tips.
Manufacturer:  Thermo Scientific™ 7442
 View more versions of this product
Catalog No. 14-387-198
Add to cart
Description
Description
Choose from a comprehensive selection of pipette tips. Thermo Scientific™ Matrix Pipette Tips are supplied in innovative new Thermo Scientific™ Flo-Thru™ racks. Matrix pipetters and pipet tips are designed together to ensure that a perfect balance exists between security of the seal and ease of application/ejection. Manufactured under tight QC specifications, these standard tips rival special low-retention tips currently on the market after pipetting fluids such as dH2O, 50% Glycerol, Hybridized Buffer and 0.1% SDS.
Benefits:
  • Sealing rings on tips ensure confident, leak-free seal without banging tips on pipetter.
  • Because the sealing rings compress, these tips also offer a secure seal on most other pipetters on the market.
  • Highly polished molds and rigorous maintenance procedures ensure low liquid retention tips without special treatments or additives.
  • Stabilization rings in racks hold tips steady when seating on pipetter.
  • Venting features in rack ensure proper heat flow during autoclaving and reduce condensation on tips.
  • Finger notch on racks allows easy removal/insertion of refill trays.
Innovative Racks and Packaging:
  • Rigid rack design ensures solid, even tip seating; rack will not flex or bend when trying to apply tips.
  • Unique Thermo Scientific™ Flo-Thru™ rack individually supports tips to prevent them from wobbling when trying to seat them on the pipetter.
  • Hinge prevents sterile lid from coming in contact with lab bench.
  • Venting along the edge or around each tip improves heat flow during autoclaving; reduces condensation that can form on the tips.
  • Dual-position lid can be propped open to prevent vacuum deformation during autoclaving.
  • Sterile racks are sold in individual bags with quick-tear openings; each rack remains sterile until ready for use.
  • Each rack is labeled with product details including re-ordering information, description, volume range, and lot number.
  • Racks are constructed of clean recycled materials and are made as small as possible to reduce waste.
  • Racks are marked with appropriate symbols to support recycling initiatives.
Specifications
Specifications
Thermo Scientific™ Matrix Pipettor
Matrix 125, 384, Hinged
Non-Filtered
Natural
DNase, RNase, DNA, ATP, PCR Inhibitor, and Pyrogen Free
Matrix
Rack
125μL
Sterile
5.3cm
Thermo Scientific™ Matrix Pipettor