Learn More
Thermo Scientific™ Matrix™ Reagent Reservoirs
Accomodate a variety of applications with these uniquely designed, innovative reagent reservoirs.
Manufacturer:  Thermo Scientific™ 8076
 View more versions of this product
Catalog No. 14-387-066
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Accomodate a variety of applications with uniquely designed, innovative Thermo Scientific™ Matrix Reagent Reservoirs. These sterile reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills. The 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizing recovery if using only a single channel and a small amount of reagent.
  • Trough within a trough maximizes amount of liquid accessible to pipet tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Sterile reservoirs are packaged in snap and tear bags, maintaining sterility while being easy to open
Description & Specifications
Specifications
No
2.5 oz.
PP Lid; Fits 75mL; 10 per case
10 per case
10
White
Reservoir Lids
75mL
Polypropylene
10
2.5 oz., 75mL
Reservoir