Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips) Available on GSA/VA Contract for Federal Government customers only.
Achieve ultimate performance in automated pipetting with Thermo Scientific™ D.A.R.T.'s™.
Manufacturer: Thermo Scientific™ 5311
 View more versions of this product
Catalog No. 14-387-679
  Check Availability
Add to Cart
    Specifications
    Description 384 Tips, Requires special aluminum tip magazine
    Volume (Metric) 30μL
    Filtered No
    Sterility Nonsterile
    Packaging 10 x 384
    For Use With (Equipment) Matrix SerialMate Pipetter
    Product Line Matrix SerialMate
    Achieve ultimate performance in automated pipetting with Thermo Scientific™ D.A.R.T.'s™. Unlike conventional automation tips, D.A.R.T.'s are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision. Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x3, PlateMate™ Plus Systems and Versette.
    Features:
    • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access.
    • Face seal design ensures consistent tip-to-tip height for precise low volume dispensing
    • Highly polished molds and cores ensure low liquid retention.
    • Small internal tip orifice provides better small volume droplet formation, resulting in optimal accuracy and precision.
    • Sterile, sterile filtered, wide-bore, and extended-length tip options are available
    • Tips are thoroughly inspected for proper formation and straightness
    • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing.
    • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination.
    Includes: 20 magazines per case
    Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x3 and PlateMate™ Plus Systems.
  • Description & Specifications
Related Products
Product Content Feedback