Thermo Scientific™ Matrix™ TallTip™ Extended Length Pipette Tips Available on GSA/VA Contract for Federal Government customers only.

Maximize reach with Thermo Scientific™ Matrix™ TallTip™ Extended Length Pipette Tips, which allow access to the bottom of test tubes, bottles and flasks.

Manufacturer: Thermo Scientific™ 7281

 View more versions of this product

Catalog No. 14-387-151

  Check Availability
Add to Cart

    Specifications

    Packaging 10 x 96/Rack
    For Use With (Equipment) Universal Fit
    Volume (Metric) 250μL
    Product Line Matrix TallTip
    Filtered No
    Tip Style Extended Length Tip
    Sterility Nonsterile

    Maximize reach with Thermo Scientific™ Matrix™ TallTip™ Extended Length Pipette Tips, which allow access to the bottom of test tubes, reagent bottles, flasks and other vessels without touching the barrel of the pipetter against the side of the tube. Nonbeveled, narrow tip ends provide cleaner touch-off of sidewalls for greater precision.

    Benefits:

    • Prevent cross-contamination from vessel walls
    • Allow the use of large volume tips in small spaces resulting in less liquid displacement
    • 102mm maximum length offers safe access to 100mm test tubes
    • Thermo Scientific™ Flo-Thru™ rack reduces tip wobble and improves airflow while autoclaving
    • Universal fit works with most major brands of pipetters

    Matrix Pipet Tip Features

    • Sealing rings on tips ensure a confident seal without banging tips on pipetter
    • Highly polished molds and rigorous maintenance procedures ensure low liquid retention tips without special treatments or additives
    • Stabilization rings in racks hold tips steady when seated on a pipetter
    • Rigid rack design ensures solid, even tip seating
    • Venting features in rack ensure proper heat flow during autoclaving and reduce condensation on tips
    • Hinged lid prevents sterile lid from coming in contact with lab bench
    • Finger notch on racks allows easy removal/insert of refill trays
    • Snap-and-tear bags make it easy to open each individually wrapped sterile rack
    • Well-labeled racks simplify record keeping, tip identification and re-ordering
    • Environmentally sensitive reduced rack size reduces waste
    • Racks utilize recycled material and are marked with appropriate symbols to support recycling initiatives

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit