Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Matrix™ TallTip™ Extended Length Pipette Tips

Maximize reach with Thermo Scientific™ Matrix™ TallTip™ Pipette Tips, which allow access to the bottom of test tubes, bottles and flasks.

$53.60 - $139.60

Specifications

For Use With (Equipment) Universal Fit Pipettor
Product Line Matrix TallTip
Filtered Non-filtered
View More Specs

Products 7
Catalog Number Mfr. No. Sterility Tip Style Volume (Metric) Format Price Quantity & Availability  
Catalog Number Mfr. No. Sterility Tip Style Volume (Metric) Format Price Quantity & Availability  
14-387-154
View Documents
Thermo Scientific™
7080
Non-sterile Extended Length 300 μL Bulk, 500 Tips
Case of 500 for $53.60
Add to cart
 
14-387-169
View Documents
Thermo Scientific™
7151
Non-sterile Universal Fit 250 μL Rack, 10 x 96 Tips
Case of 960 for $119.90
Add to cart
 
14-387-168
View Documents
Thermo Scientific™
7250
Non-sterile Extended Length 250 μL Bulk, 1000 Tips
Case of 1000 for $90.60
Add to cart
 
14-387-150
View Documents
Thermo Scientific™
7280
Non-sterile Extended Length 250 μL Bulk, 500 Tips
Case of 500 for $53.60
Add to cart
 
14-387-151
View Documents
Thermo Scientific™
7281
Non-sterile Extended Length 250 μL Rack, 10 x 96 Tips
Case of 960 for $128.30
Add to cart
 
14-387-147
View Documents
Thermo Scientific™
7631
Non-sterile Extended Length 30 μL Rack, 10 x 96 Tips
Case of 960 for $118.10
Add to cart
 
14-387-148
View Documents
Thermo Scientific™
7632
Sterile Extended Length 30 μL Rack, 10 x 96 Tips
Case of 960 for $139.60
Add to cart
 
Description

Description

Maximize reach with Thermo Scientific™ Matrix™ TallTip™ Pipette Tips, which allow access to the bottom of test tubes, reagent bottles, flasks and other vessels without touching the barrel of the pipetter against the side of the tube. Nonbeveled, narrow tip ends provide cleaner touch-off of sidewalls for greater precision.

Benefits:
  • Prevent cross-contamination from vessel walls
  • Allow the use of large volume tips in small spaces resulting in less liquid displacement
  • 102mm maximum length offers safe access to 100mm test tubes
  • Thermo Scientific™ Flo-Thru™ rack reduces tip wobble and improves airflow while autoclaving
  • Universal fit works with most major brands of pipetters

Matrix Pipet Tip Features

  • Sealing rings on tips ensure a confident seal without banging tips on pipetter
  • Highly polished molds and rigorous maintenance procedures ensure low liquid retention tips without special treatments or additives
  • Stabilization rings in racks hold tips steady when seated on a pipetter
  • Rigid rack design ensures solid, even tip seating
  • Venting features in rack ensure proper heat flow during autoclaving and reduce condensation on tips
  • Hinged lid prevents sterile lid from coming in contact with lab bench
  • Finger notch on racks allows easy removal/insert of refill trays
  • Snap-and-tear bags make it easy to open each individually wrapped sterile rack
  • Well-labeled racks simplify record keeping, tip identification and re-ordering
  • Environmentally sensitive reduced rack size reduces waste
  • Racks utilize recycled material and are marked with appropriate symbols to support recycling initiatives
Specifications

Specifications

Universal Fit Pipettor
Matrix TallTip
Non-filtered
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit