Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAVS, Mouse anti-Human, Clone: Adri-1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP280052

Catalog No. NB013180


Add to cart

Description

Description

MAVS Monoclonal antibody specifically detects MAVS in Human Samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

MAVS
Adri-1
Unconjugated
PBS containing 10% glycerol
CARD adapter inducing interferon beta, CARD adapter inducing interferon-beta, CARD adaptor inducing IFN-beta, CARDIF, DKFZp547C224, DKFZp666M015, FLJ27482, IFN-B promoter stimulator 1, Interferon beta promoter stimulator protein 1, interferon-beta promoter stimulator protein 1, IPS-1FLJ41962, IPS1MGC3260, KIAA1271FLJ35386, mitochondrial antiviral signaling protein, mitochondrial antiviral-signaling protein, mitochondrial viral signaling protein, Putative NF-kappa-B-activating protein 031N, virus-induced signaling adapter variant 1b, virus-induced signaling adaptor variant 1a, Virus-induced-signaling adapter, VISAFLJ38051
Mouse
IgG2b
100 ug
Immune Dysfunction, Immune System Diseases, Immunology, Protein Kinase
Primary
57506
Human
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
1 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Purified
MAVS
Recombinant human MAVS(aa 160-450).
Protein G purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Recognizes human MAVS.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit