Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Maxima™ H Minus cDNA Synthesis Master Mix
Provide one-tube master mix for convenient and consistent cDNA synthesis for two-step RT-qPCR applications. The format helps streamline the RT-qPCR workflow, saves time, reduces pipeting errors to give customers sensitive, reliable and fast cDNA synthesis.
Manufacturer: Thermo Scientific™ M1662
Description
Thermo Scientific Maxima H Minus Reverse Transcriptase (RT), an engineered RNase H minus enzyme, is formulated into convenient one-tube master mix with a simplified protocol, enabling consistency and maximum control of RT-qPCR. It was developed through molecular evolution to improve thermostability, robustness and synthesis rates. Maxima H Minus RT also shows up to a 50x increase in processivity, has no RNase H activity, and demonstrates higher sensitivity.
- Efficiently transcribes large and small amounts of RNA
- Quick and convenient setup eliminates pipeting caused variability
- Reduces risk of cross-contamination between samples
- Consistently higher sensitivity for wide variety of gene targets
- High linearity in cDNA synthesis over a broad dynamic range
- Complete enzyme activity from 42° to 55°C
Specifications
qPCR | |
Tube | |
5X | |
800 μL | |
Dry Ice | |
RNA | |
SuperMix or Master Mix | |
50°C inculbation temperature for RT reactions |
H Minus cDNA Synthesis Master Mix Kit | |
200 Reactions | |
cDNA Synthesis Master Mix | |
Maxima&trade | |
200 | |
Real-Time PCR | |
Maxima H Minus |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title