Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Maxima™ H Minus cDNA Synthesis Master Mix, with dsDNase

Provide one-tube master mix for convenient and consistent cDNA synthesis for two-step RT-qPCR applications. The format helps streamline the RT-qPCR workflow, saves time, reduces pipeting errors to give customers sensitive, reliable and fast cDNA synthesis.

$437.00 - $1404.00

Specifications

Concentration 5X (Maxima H Minus cDNA Synthesis Master Mix), 10X (dsDNase Buffer)
Final Product Type cDNA (First-Strand)
Format Tube
For Use With (Application) Real-Time PCR
PCR Method RT-PCR
View More Specs

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
FERM1681
View Documents
Thermo Scientific™
M1681
50 Reactions
Each for $437.00
Add to cart
 
FERM1682
View Documents
Thermo Scientific™
M1682
200 Reactions
Each for $1,404.00
Add to cart
 
Description

Description

Thermo Scientific Maxima H Minus Reverse Transcriptase (RT), an engineered RNase H minus enzyme, is formulated into convenient one-tube master mix with a simplified protocol, enabling consistency and maximum control of RT-qPCR. It was developed through molecular evolution to improve thermostability, robustness and synthesis rates. Maxima H Minus RT also shows up to a 50x increase in processivity, has no RNase H activity, and demonstrates higher sensitivity.
  • Efficiently transcribes large and small amounts of RNA
  • Quick and convenient setup eliminates pipeting caused variability
  • Reduces risk of cross-contamination between samples
  • Consistently higher sensitivity for wide variety of gene targets
  • High linearity in cDNA synthesis over a broad dynamic range
  • dsDNase for fast and more efficient gDNA removal in RNA prep and no-RT control for RT-qPCR
  • dsDNase treated RNA samples show no decrease in RNA integrity or quantity
  • Complete enzyme activity from 42° to 55°C
Specifications

Specifications

5X (Maxima H Minus cDNA Synthesis Master Mix), 10X (dsDNase Buffer)
Tube
RT-PCR
Reverse Transcription
Up to 20 kb
Reverse Transcription
50°C inculbation temperature for RT reactions
cDNA Synthesis Master Mix
Dry Ice
cDNA (First-Strand)
Real-Time PCR
Master Mix
RNA
RNA
H Minus cDNA Synthesis Master Mix Kit
Maxima™
Maxima H Minus
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit