Learn More
Thermo Scientific™ Maxima H Minus First Strand cDNA Synthesis Kits Promotion
Synthesize long cDNAs up to 20 kb at elevated temperatures (up to 65°C) with this complete system for highly efficient synthesis of first strand cDNA
Manufacturer:  Thermo Scientific™ K1682
 View more versions of this product
Catalog No. FERK1682PM
Add to cart
Description
Description
  • Increased reaction temperatures -the first strand of cDNA can be synthesized within the 42 to 65°C temperature range
  • High yields of full-length first strand cDNA -with RNA templates up to 20 kb
  • Flexible priming -oligo(dT)18, random hexamer, or gene-specific primers
Specifications
Specifications
First Strand cDNA synthesis for RT-PCR, Construction of cDNA libraries, Generation of probes for hybridization, Antisense RNA synthesis
100 x 20μL Reactions