Learn More
Thermo Scientific™ Maxima H Minus Reverse Transcriptase Promotion
A highly thermostable, high processivity reverse transcriptase lacking RNase H activity
Manufacturer:  Thermo Scientific™ EP0753
 View more versions of this product
Catalog No. FEREP0753PM
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
-20°C
First strand cDNA synthesis for RT-PCR and real-time RT-PCR, reverse transcription at elevated temperatures to reduce effects of secondary structure, synthesis of cDNA for cloning and expression, generation of labeled cDNA probes for microarrays
5X RT (250mM Tris-HCl buffer (pH 8.3 at 25°C), 375mM KCl, 15mM MgCl2, 50mM DTT)
4 x 10000U